Learn More
STEMCELL Technologies StemSpan™ SFEM II
Product Code. 15764682
Encompass
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15764682 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15764682 Supplier STEMCELL Technologies Supplier No. 09605
Please to purchase this item. Need a web account? Register with us today!
Serum-free medium for culture and expansion of hematopoietic cells.
StemSpan™ Serum-Free Expansion Medium II (SFEM II) is a modified version of StemSpan SFEM. It has been developed for the in vitro culture and expansion of human hematopoietic cells. This medium contains pre-tested bovine serum albumin, insulin, transferrin, and other supplements in Iscove's MDM. Recombinant hematopoietic growth factors, required for the optimal growth and expansion of hematopoietic cells, have not been added to StemSpan SFEM II. This allows users the flexibility to prepare medium that meets their requirements.
Using appropriate cytokines (e.g. StemSpan CC100, StemSpan CC110, or StemSpan CD34+ Expansion Supplement), StemSpan SFEM II can be used for the expansion of total nucleated cells and CD34+ cells from cord blood, bone marrow, or other cell sources. StemSpan SFEM II can also be used to expand and differentiate lineage-committed progenitor cells to generate erythroblasts, granulocytes, monocytes, or megakaryocytes when used with StemSpan Erythroid Expansion Supplement (02692), StemSpan Myeloid Expansion Supplement (02693), StemSpan Myeloid Expansion Supplement II (02694), or StemSpan Megakaryocyte Expansion Supplement (02696), respectively. 
Specifications
Cell Type Hematopoietic Stem and Progenitor Cells
Product Type SFEM II
For Use With (Application) Cell Culture, Expansion
Quantity 100 mL
Species Human