missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies StemSpan™ Erythroid Expansion Supplement (100X)
Description
- StemSpan™ Erythroid Expansion Supplement (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of erythroid progenitor cells from CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples
- When added to serum-free medium, StemSpan™ Erythroid Expansion Supplement typically promotes the production of thousands of erythroid cells per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells
- See data tab for more details
- StemSpan™ Erythroid Expansion Supplement (100X) is intended for use in combination with any of the following StemSpan™ media: StemSpan™ SFEM (Catalog #09600)
- StemSpan™ SFEM II (Catalog #09605)
- StemSpan™-XF (Catalog #100-0073)
- StemSpan™-ACF Erythroid Expansion Medium (Catalog #09860)
Specifications
Specifications
| Cell Type | Erythroid Cells, Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Differentiation, Expansion |
| Quantity | 1 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?