missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies StemSpan™ CC100

Product Code. 17158051
Encompass
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17158051 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17158051 Supplier STEMCELL Technologies Supplier No. 02690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serum-free culture supplement for expansion of human hematopoietic cells

  • StemSpan™ CC100 cytokine cocktail contains a combination of both early- and late-acting recombinant human (rh) cytokines formulated to support the proliferation of human hematopoietic cells
  • It is supplied as a 100X concentrate
  • When added to serum-free medium, StemSpan™ CC100 promotes the expansion of CD34+ cells isolated from human cord blood and bone marrow, and is recommended if high cell yields and large numbers of CD34+ cells are desired
  • We recommend using StemSpan™ CC100 in combination with any of the following StemSpan™ media: StemSpan™ SFEM (Catalog #09600)
  • StemSpan™ SFEM II (Catalog #09605)
  • StemSpan™-XF (Catalog #100-0073)
  • StemSpan™-ACF Without Phenol Red (Catalog #100-0130)

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Expansion
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.