missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Neural Crest Differentiation Kit
Description
- STEMdiff™ Neural Crest Differentiation Kit creates a serum-free medium for differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to neural crest cells
- These neural crest cells, which are characterized by neural crest markers such as SOX10 and CD271, can be differentiated to several downstream derivatives including chondrocytes, osteoblasts, and peripheral neurons
- This medium is compatible with human ES and iPS cells maintained in either mTeSR™1 (Catalog #85850), mTeSR™ Plus (Catalog #05825) or TeSR™-E8™ (Catalog #05990)
Specifications
Specifications
| Cell Type | Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Neurons, Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Differentiation, Toxicity Assay |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?