missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Forebrain Neuron Maturation Kit
Description
- The STEMdiff™ Forebrain Neuron Maturation Kit is used in conjunction with the STEMdiff™ Forebrain Neuron Differentiation Kit (Catalog #08600) to generate a mixed population of forebrain-type (FOXG1-positive) neurons from neural progenitor cells derived from human pluripotent stem cells
- The neurons derived are highly pure (≥ 90% class III β-tubulin-positive neurons; < 10% GFAP-positive astrocytes), functional and can be maintained long-term in culture
- Neurons derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation
Specifications
Specifications
| Cell Type | Neural Cells, PSC-Derived, Neurons |
| For Use With (Application) | Cell Culture, Characterization, Differentiation, Functional Assay, Immunocytochemistry, Phenotyping |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?