missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Definitive Endoderm Kit

Product Code. 13449008
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13449008 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13449008 Supplier STEMCELL Technologies Supplier No. 05110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Defined animal component-free medium for the differentiation of human ES and iPS cells to definitive endoderm

  • STEMdiff™ Definitive Endoderm Kit is a complete, serum- and animal component-free medium and supplement kit that supports highly efficient differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to definitive endoderm cells
  • Cells differentiated using STEMdiff™ Definitive Endoderm Kit express high levels of endoderm markers, including CD184 (CXCR4), SOX17, FOXA2, and c-KIT, and lack expression of ectoderm, mesoderm, and pluripotency markers
  • The definitive endoderm cells produced using this kit are multipotent and capable of further differentiation towards cells of the pancreatic, intestinal, pulmonary, and hepatic lineages, thus providing a robust tool for developmental studies, disease modeling, and drug discovery
  • ​This kit is optimized for differentiation of cells maintained in mTeSR™1
  • For differentiation of cells maintained in TeSR™-E8™, please see the STEMDiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimized)

Specifications

Cell Type Airway Cells, Endoderm, PSC-Derived, Hepatic Cells, Intestinal Cells, Pancreatic Cells, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.