Learn More
Nabertherm Steel Collecting Pan
For use with LE 2 furnace. Protects the furnace and provides easy loading.
Brand: Nabertherm 691400626
Additional Details : Weight : 5.00000kg
Description
- Steel construction
- For use with LE 2 model furnace
- Dimensions (L x W x H): 110 x 170 x 20 mm
- Maximum Temperature: 1100°C
Specifications
Protects the furnace and provides easy loading |