Learn More
Stearylpyridinium Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Stearylpyridinium Chloride |
| CAS | 3165-81-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23 H42 N . Cl |
| Quantity | 1 g |
| Synonym | Pyridinium','1-octadecyl-','chloride (1:1), 1-Octadecylpyridinium chloride (6CI,7CI), Pyridinium','1-octadecyl-','chloride (8CI,9CI), Octadecylpyridinium chloride, Stearylpyridinium chloride |
| InChI Formula | InChI=1S/C23H42N.ClH/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-18-21-24-22-19-17-20-23-24;/h17,19-20,22-23H,2-16,18,21H2,1H3;1H/q+1;/p-1 |
| SMILES | [Cl-].CCCCCCCCCCCCCCCCCC[n+]1ccccc1 |
| IUPAC Name | 1-octadecylpyridin-1-ium;chloride |
| Molecular Weight (g/mol) | 368.039 |
| Show More |