Learn More
STC1 Monoclonal Antibody (OTI1E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809955S
Description
STC1 Monoclonal antibody specifically detects STC1 in Human samples. It is validated for Western BlotSpecifications
| STC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96NU7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 144193 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1E9 | |
| Unconjugated | |
| AMDHD1 | |
| stanniocalcin; stanniocalcin 1; stanniocalcin-1; STC; STC1; STC-1 | |
| Full length human recombinant protein of human AMDHD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.