missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Stathmin-2/STMN2 Polyclonal specifically detects Stathmin-2/STMN2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, In vivo assay, Knockdown Validated.
Specifications
Specifications
| Antigen | Stathmin-2/STMN2 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Frozen), In vitro Assay, Immunohistochemistry, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:4000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:4000, Immunohistochemistry-Paraffin reported in scientific literature (PMID 31182472), Immunohistochemistry-Frozen 1:1000, In vitro assay reported in scientific literature (PMID 22726832), In vivo assay reported in scientific literature (PMID 22726832), Immunohistochemistry Whole-Mount reported in scientific literature (PMID 35042776), Knockdown Validated |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Alias | FLJ34868, FLJ50995, neuron-specific growth-associated protein, Protein SCG10, SCG10, SCG10neuronal growth-associated protein (silencer element), SCGN10stathmin-2, stathmin-like 2, superior cervical ganglia, neural specific 10, Superior cervical ganglion-10 protein |
| Gene Symbols | STMN2 |
| Host Species | Rabbit |
| Immunogen | C-terminal peptide of mouse STMN2. [Swiss-Prot P55821] |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?