missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Stathmin-2/STMN2 Antibody [HRP], Novus Biologicals™

Product Code. 30034192
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30034192 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30034192 Supplier Novus Biologicals™ Supplier No. NBP149461H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Stathmin-2/STMN2 Polyclonal antibody specifically detects Stathmin-2/STMN2 in Bovine,Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),In vitro Assay,In vivo Assay,Immunohistochemistry (Whole-Mount),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stathmin-2/STMN2
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, In vivo Assay, Immunohistochemistry (Whole-Mount), KnockDown, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias FLJ34868, FLJ50995, neuron-specific growth-associated protein, Protein SCG10, SCG10, SCG10neuronal growth-associated protein (silencer element), SCGN10stathmin-2, stathmin-like 2, superior cervical ganglia, neural specific 10, Superior cervical ganglion-10 protein
Host Species Rabbit
Immunogen C-terminal peptide of mouse STMN2. [Swiss-Prot P55821]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11075
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.