missing translation for 'onlineSavingsMsg'
Learn More

STAT5-IN-1, MedChemExpress

Product Code. p-200096061
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494696 100 mg 100mg
16493608 10 mg 10mg
16482786 25 mg 25mg
16466337 5 mg 5mg
16430007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494696 Supplier Medchem Express Supplier No. HY101853

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress STAT5-IN-1 is a STAT5 inhibitor with an IC50 of 47 μM for STAT5β isoform.

STAT5-IN-1 is a STAT5 inhibitor with an IC50 of 47 μM for STAT5β isoform.

Especificaciones

Chemical Name or Material STAT5-IN-1
CAS 285986-31-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H11N3O3
Quantity 100 mg
Solubility Information DMSO : 20 mg/mL (68.19 mM; Need ultrasonic)
SMILES O=C(C1=CC=CN=C1)N/N=C/C2=COC3=CC=CC=C3C2=O
Molecular Weight (g/mol) 293.28
Formula Weight 293.28
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.