Learn More
STAT5-IN-1, MedChemExpress
Product Code. 16494696
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494696 100 mg 100mg
16493608 10 mg 10mg
16482786 25 mg 25mg
16466337 5 mg 5mg
16430007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16494696 Supplier Medchem Express Supplier No. HY101853
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress STAT5-IN-1 is a STAT5 inhibitor with an IC50 of 47 μM for STAT5β isoform.
STAT5-IN-1 is a STAT5 inhibitor with an IC50 of 47 μM for STAT5β isoform.
Specifications
Chemical Name or Material STAT5-IN-1
CAS 285986-31-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H11N3O3
Quantity 100 mg
Solubility Information DMSO : 20 mg/mL (68.19 mM; Need ultrasonic)
SMILES O=C(C1=CC=CN=C1)N/N=C/C2=COC3=CC=CC=C3C2=O
Molecular Weight (g/mol) 293.28
Formula Weight 293.28
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease