missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stargazin Antibody (S245-36), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22411F
This item is not returnable.
View return policy
Description
Stargazin Monoclonal specifically detects Stargazin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Stargazin | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| CACNG2 | |
| Fusion protein amino acids 203-323 (Cytoplasmic C-terminus) of rat TARPGamma2 | |
| 0.1 mL | |
| Primary | |
| Detects approx 35-55kDa. Does not cross-react with TARPGamma3. | |
| Store at 4C in the dark. | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence | |
| S245-36 | |
| Immunocytochemistry/Immunofluorescence | |
| calcium channel, voltage-dependent, gamma subunit 2, FLJ41437, MGC138502, MGC138504, Neuronal voltage-gated calcium channel gamma-2 subunit, stargazin, voltage-dependent calcium channel gamma-2 subunit | |
| Mouse | |
| Protein G purified | |
| Neuroscience | |
| 10369 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction