Learn More
Thermo Scientific™ 2'-OMe-PAC-A Phosphoramidite, standard grade, serum vial bottle
Shop All Thermo Scientific ProductsDescription
Ensure quality and reliability for RNA oligonucleotide synthesis with Thermo Scientific™ Standard RNA 2'-TBDMS, 2'-OMe and 2'-F Phosphoramidites.
2'-OMe-PAC-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20-mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N6-phenoxyacetyl-2'-O-methyl-adenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C49H56N7O9P
• Base: A
• Base Protection: PAC
• Formula weight: 918.0 g/mol
• CAS number: 128219-82-9
• HPLC purity: ≥98.0%
• 31P NMR purity: ≥98.0%
Specifications
Specifications
| For Use With (Equipment) | AKTA Synthesizers, Applied Biosystems Synthesizers |
| Product Type | 2'-OMe-PAC-A CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Adenosine (A) |
| Reagent Features | 2'-OMe modified, Fast deprotection |
| Shipping Condition | Room Temperature |
| Purity | 98% |
| Quantity | 1 g |
| Reagent Type | Phosphoramidite |
| Grade | Standard |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.