Learn More
Tenak™ Standard Rack for Microtiterplates, for Chest Freezers, 65 mm Shelf Height
Stainlesss Steel Standard Rack for chest freezers is designed to hold microtiterplates.
Brand: Tenak™ TE27194
Additional Details : Weight : 0.75000kg
Description
- For use in chest freezers
- Racks come with a locking rod to secure the cryoboxes
- Constructed of stainless steel
Specifications
Locking rod | |
Stainless Steel | |
135 x 92 x 403 mm |
Microtiterplates (6) | |
Chest Frseezers | |
65 mm |