Learn More
Fritsch™ Standard Quality Agate Mortar With Pestle
Specifications
Specifications
| Capacity (Metric) | 25 mL |
| Diameter (Metric) Inner | 60mm |
| Diameter (Metric) Outer | 75mm |
| Product Type | Agate Mortar |