Learn More
Ohaus™ Stainless Steel Weight, OIML F1, OH
Product Code. 15673384 Shop All Ohaus Products
Click to view available options
Weight (Metric):
100 g
200 g
1 kg
Unit Size:
Each
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15673384
Brand: Ohaus™ 11117041
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Single body design weight adjusted to meet or exceed 0IML F1 tolerances
  • Stainless steel construction
  • Include a weight case
  • Single body design without adjusting cavity
TRUSTED_SUSTAINABILITY
Specifications
Description Weight, 100g, OIML F1, OH
Classification OIML F1 Weights
Material Stainless Steel
Weight (Metric) 100 g
Tolerance Meets or exceeds OIML F1