Learn More
Linker Industrietechnik™ Stainless Steel Test Sieve, 200 mm OD, 50 mm H, Soft Solder Seam
Product Code. 10045100
Click to view available options
Pore Size:
63 μm
125 μm
250 μm
Unit Size:
Each
Product Code. 10045100
Brand: Linker Industrietechnik™ 552050222038
Only null left
Add to basket
Add to basket
Constructed with a soft solder seam and a sieve of stainless steel cloth.
  • Stainless steel ring construction
  • Stainless steel sieve cloth/skin
  • Soft solder seam
  • Heat resistant
  • Available in a variety of sieve mesh sizes
Specifications
Description Heat resistant up to 170°C
Pore Size 250 μm
Certifications/Compliance 2.1 EN 10 204, ISO 3310-1
Height (Metric) Internal: 50 mm
U.S. Standard No. 60
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Screen Size 60 Mesh
Mesh Material Stainless Steel
Sieve Number 60