Learn More
Horst™ Stainless Steel Temperature Probe
Product Code. 10087071
Click to view available options
Depth (Metric):
75 mm
Unit Size:
Each
Product Code. 10087071
Brand: Horst™ 70202
Only null left
Add to basket
Add to basket
Specifications
Depth (Metric) 75 mm
Diameter (Metric) Probe 1.5 mm
Height (Metric) 145 mm
Length (Metric) Cable 1.5 m
Length (Metric) Probe 250 mm
Product Type Temperature Probe
Max. Temperature (Metric) 999°C
Min. Temperature (Metric) 20°C
Temperature Range (Metric) 20°C to 999°C
Accuracy 0.01