Learn More
DWK Life Sciences Kimble™ Ultra-Ware™ Stainless Steel Support Screen
The Kimble™ Ultra-Ware™ Stainless Steel Support Screen is suitable for use with Kimble™ Ultra-Ware™ Microfiltration Assemblies.
Brand: DWK Life Sciences 953711-0000
Additional Details : Weight : 0.80000kg
Description
- Product Type: Support Screen
- Material: 304 Stainless Steel
- Size: 25 mm
- Mesh Type: 80 × 80 Wire Mesh [wire diameter: 0.18 mm (0.007 in.)]
- Opening size: 0.24 mm (0.0095 in.)
- Wire diameter: 0.18 mm (0.007 in.)
- Open Area: 19.4%
- For Use with: Kimble™Ultra-Ware™ Microfiltration Assembly
Specifications
Not Autoclavable | |
Wire Mesh | |
Stainless Steel (304) | |
Non-sterile | |
Round | |
25 mm |
Reusable | |
Kimble Ultra-Ware Microfiltration Assembly | |
Stainless Steel | |
Open Area: 19.4% | |
Support Screen |