Learn More
KGW Isotherm™ Stainless Steel Stirrer Coupling
Product Code. 10005201
Click to view available options
Type:
Stirrer Coupling
Unit Size:
Each
Product Code. 10005201
Brand: KGW Isotherm™ 40470
This item requires a price update prior to being ordered.
Request a Price for this item here.
A flexible coupling solution for stirrers in the laboratory. KGW Isotherm™ Stainless Steel Stirrer Couplings are for shafts from 8 to 10mm in diameter.
  • Sturdy construction
  • Supplied in packs of one
Specifications
For Use With (Application) Compensate Any Vibrations Between Stirrer and Tools
Includes Metal Spring
Quantity 1 Pack
Type Stirrer Coupling