missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Stainless Steel Rod

Product Code. 15752349 Shop All Ohaus Products
Change view
Click to view available options
Length (Metric):
40 mm
50 mm
150 mm
300 mm
460 mm
610 mm
910 mm
1220 mm
1520 mm
1830 mm
2440 mm
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15752349 1520 mm Each
15772339 40 mm Each
15782339 50 mm Each
15792339 150 mm Each
15702349 300 mm Each
15712349 460 mm Each
15722349 610 mm Each
15732349 910 mm Each
15742349 1220 mm Each
15762349 1830 mm Each
15772349 2440 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 15752349 Supplier OHAUS™ Supplier No. 30392301
Add to basket
Add to basket
This item is not returnable. View return policy

OHAUS™ Stainless Steel Rod provides secure support when used with an OHAUS™ Lab Frame.

  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Dependable connectors secure rods and clamps to meet your requirements and five unique holders in a variety of sizes and materials accommodate different rods.
TRUSTED_SUSTAINABILITY

Specifications

Type Lab Frame Rod
Material Stainless Steel
Description Clamp, Rod, Steel 152 cm, CLR-RODS152
Diameter (Metric) 13 mm
Length (Metric) 1520 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.