Learn More
Endecotts™ Stainless Steel Receiver
Product Code. 12934225
Change view
Click to view available options
Diameter (Metric):
100 mm
200 mm
230 mm
300 mm
450 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12934225 100 mm Each
11537524 200 mm Each
10736935 300 mm Each
12825691 450 mm Each
10566734 450 mm Each
10492065 230 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12934225 Supplier Endecotts™ Supplier No. 100S/STREC
  • ONLINE EXCLUSIVE PRICE
    80.66 €
    36.69 € / Pack of 1
    Save 43.97 €
    54% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
Full height receivers for use with test sieves. Endecotts™ Stainless Steel Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis.
  • Stainless steel construction
  • For use with Stainless Steel Woven Wire Mesh Test Sieves
Specifications
Product Type Receiver
Material Stainless Steel
Diameter (Metric) 100 mm
For Use With (Application) For sieves
Packaging Quantity 1 Ea.