Learn More
Bochem™ Stainless Steel Petri Dish
Product Code. p-7146775
Click to view available options
Diameter (Metric):
60 mm
90 mm
120 mm
Unit Size:
Each
Product Code. 15861677
Brand: Bochem™ 8635
Only null left
Add to basket
Add to basket
Bochem™ Stainless Steel Petri Dish includes a Stainless Steel lid.
  • Stainless Steel
  • Lid included
Specifications
Closure Included Closure Included
Material Stainless Steel
Shape Round
Height (Metric) 20 mm
Diameter (Metric) 60 mm