Learn More
SPEX CertiPrep™ Stainless Steel Long Cryogenic Transfer Hose
Product Code. 15350502 Shop All SPEX Certiprep Products
Change view
Click to view available options
Length (Metric):
1.8 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15350502 1.8 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15350502 Supplier SPEX CertiPrep™ Supplier No. 6907
Please to purchase this item. Need a web account? Register with us today!
Useful for transferring cryogenic fluids such as liquid nitrogen.
  • Flexible stainless steel construction.
  • Withstands liquid nitrogen temperatures.
  • Fitted with a 3/8 in. (9.5 mm) NPT male fitting on one end and aCGA295 female fitting on the other end.
  • Compatible with 6905 Portable Cryogenic Dewar and 6820 Auto-Fill System.
Specifications
For Use With (Equipment) 6905 Portable Cryogenic Dewar and 6820 Auto-Fill System.
Length (English) 6 ft.
Length (Metric) 1.8 m