Learn More
asecos™ Stainless Steel Immersion Container
Shop All asecos Products
Click to view available options
Capacity (Metric):
10 L
20 L
45 L
Unit Size:
Each
Description
- Stainless steel immersion basin
- Basin for the immesrsion and cleaning of materials.
Specifications
Specifications
| Capacity (English) | 2.6 gal. |
| Capacity (Metric) | 10 L |
| Color | Gray |
| Material | Stainless Steel |
| Width (English) | 10.63 in. |
| Width (Metric) | 270.00 mm |
| Height (English) | 10.63 in. |
| Height (Metric) | 270.00 mm |
| Weight (English) | 8.82 lb. |
| Weight (Metric) | 4 kg |