Learn More
Alpaco™ Stainless Steel Clamps with Vertical Screws
Click to view available options
Size (Metric) Grip:
12 to 17 mm
5 to 10 mm
7 to 12 mm
9 to 14 mm
Unit Size:
Pack of 10
Description
- Reusable
- Resists pressure
- No lubricant required for assembly
- Smooth clamping to avoid tubing damage
Specifications
Specifications
| Quantity | 10/Pk. |
| Type | Hose Clamp |
| Material | Stainless Steel |
| Size (Metric) Grip | 5 to 10 mm |