Learn More
Alpaco™ Stainless Steel Clamps with Vertical Screws
Product Code. 10359522
Click to view available options
Size (Metric) Grip:
12 to 17 mm
5 to 10 mm
7 to 12 mm
9 to 14 mm
Unit Size:
Pack of 10
This item is not returnable. View return policy
Product Code. 10359522
Brand: Alpaco™ INH17
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Provides uniform tightening around tubing for a watertight seal. Alpaco™ Stainless Steel Clamps with Vertical Screws are guaranteed waterproof and hand-lock without any tools.
  • Reusable
  • Resists pressure
  • No lubricant required for assembly
  • Smooth clamping to avoid tubing damage
Specifications
Quantity 10/Pk.
Type Clamp
Material Stainless Steel
Size (Metric) Grip 12 to 17 mm