missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel gauze, 80 mesh woven from 0.127mm (0.005in) dia wire, Type 304, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 013477.RU

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11316848

  • 206.00 € / Each
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Stainless Steel gauze, Type 304
Fe:Cr:Ni; 70:19:11 wt%
30 x 60 cm
80 mesh
Woven from 0.127mm (0.005 in.) dia. wire
MFCD00148499
Insoluble in water.
Wire Cloth
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

RUO – Research Use Only