Learn More
Bochem™ Stainless Steel Funnel
Product Code. 10663551
Change view
Click to view available options
Diameter (Metric)Top:
100mm
150mm
200mm
250mm
Height (Metric):
125 mm
155 mm
180 mm
190 mm
220 mm
260 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Height (Metric) unitSize
10663551 200mm 180 mm Each
10748741 100mm 125 mm Each
10018510 150mm 190 mm Each
10231641 150mm 155 mm Each
10539671 200mm 220 mm Each
10549671 250mm 260 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10663551 Supplier Bochem™ Supplier No. 8722
Only null left
Add to basket
Add to basket
Used for a wide range of applications. Bochem™ Stainless Steel Funnel has a conical shape.
  • Handle 
  • With air canal
  • Tube conical shape
  • 18/10 Stainless steel
Specifications
Diameter (Metric)Top 200mm
Diameter (Metric) Stem 30mm
Material Stainless Steel
Height (Metric) 180 mm