Learn More
VACUUBRAND™ Stainless Steel Elbow Pieces With Small KF Flanges
Product Code. 10360552
Click to view available options
Diameter (Metric) Inside:
10/10 mm
16/16 mm
25/25 mm
40/40 mm
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 10360552
Brand: VACUUBRAND™ 20673414
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Diameter (Metric) Inside 16/16 mm
For Use With Solenoid-operated valves VV and VV C
Material Stainless Steel
Quantity 1 Each
Size 40 mm
Type Elbow Piece