Learn More
Agencinox™ Stainless Steel Dental Trays
Product Code. 11791943
Click to view available options
Width (Metric):
105 mm
145 mm
Unit Size:
Each
Product Code. 11791943
Brand: Agencinox™ 1210
Only null left
Add to basket
Add to basket
Use these trays for a variety of applications. Agencinox™ Stainless Steel Dental Trays ideal for use in medical environments.
  • Rectangular tray
  • Stainless steel construction
  • Available in a variety of sizes
Specifications
Height (Metric) 15 mm
Length (Metric) 205 mm
Material Stainless Steel
Type Dental Tray
Width (Metric) 105 mm
For Use With (Application) Used in Dentistry
Shape Rectangular