Learn More
Fisherbrand™ Stainless Steel Round Bottom Weighing Scoops
Product Code. 11543452 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
70 mm
90 mm
105 mm
120 mm
Unit Size:
Pack of 5
Product Code. 11543452
Please to purchase this item. Need a web account? Register with us today!
Get optimum results weighing and handling samples and other laboratory materials with round bottom scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Round Bottom Weighing Scoops are an ideal accessory to have if you own a weighing scale.
  • Well-suited for pharmaceutical and environmental sampling
  • Ideal for weighing and handling of samples
  • Great for weighing messy ingredients, tiny objects and portions
  • Great accessory to have if you own a weighing scale
  • Perfect for weighing anything from liquid to gems and other materials
Specifications
Shape Cylindrical
Product Type Scoop
Width (Metric) 30 mm
Dimensions (L x W) 120 x 30 mm
Material Stainless Steel
Length (Metric) 120 mm