Learn More
Gilson™ Stainless Steel Connectors
Shop All Gilson Products
Click to view available options
Diameter (Metric):
0.6 mm
0.8 mm
1.1 mm
Unit Size:
Pack of 5
Description
- Silicone (Versilic™) tubing—is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
- ISOVERSINIC™ tubing—shows excellent resistance to strong acids, oxidizing agents, and aromatic and chlorinated solvents.
- PharMed™ tubing—meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
- PVC (Tygon™) tubing—handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
Specifications
Specifications
| Diameter (Metric) | 1.1 mm |
| Material | Stainless Steel |
| For Use With | For small bore tubing |
| Packaging Quantity | 5 Set |
| Includes | Metal sleeve |
| Product Type | Connector |