Learn More
Schneider Gerd™ Laboratory Scoop
Product Code. 10716115
Click to view available options
Size:
100 x 320 mm
350 x 130 mm
Blade Material:
Cast Aluminum
Stainless Steel
Unit Size:
Each
Product Code. 10716115
Brand: Schneider Gerd™ 532
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these scoops for a variety of applications. Schneider Gerd™ Laboratory Scoops are available in cast aluminum and stainless steel.
  • Metal lab scoops
  • For scooping or weighing solids
Specifications
Size 100 x 320 mm
Blade Material Stainless Steel
Length (Metric) 100 mm