Learn More
Ohaus™ Stainless Steel Calibration Weight Weight - OIML E1 Wire, NVLAP
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E1 Tolerances
177.00 €
Specifications
Certifications/Compliance | NVLAP Accredited Certificate |
---|---|
Material | Stainless Steel (316) |
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16652302
|
Ohaus™
30517963 |
1 mg |
177.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16662302
|
Ohaus™
30517964 |
2 mg |
177.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16672302
|
Ohaus™
30517965 |
5 mg |
177.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16682302
|
Ohaus™
30517966 |
10 mg |
177.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16692302
|
Ohaus™
30517967 |
20 mg |
177.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16602312
|
Ohaus™
30517968 |
50 mg |
177.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16612312
|
Ohaus™
30517969 |
100 mg |
177.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16622312
|
Ohaus™
30517970 |
200 mg |
177.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16632312
|
Ohaus™
30517971 |
500 mg |
177.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
Specifications
NVLAP Accredited Certificate | |
Stainless Steel (316) | |
Protective Case |