Learn More
Masterflex™ Straight Fitting, Barb x NPT(M), Stainless Steel
Product Code. 11709659
Click to view available options
Tubing Size:
Fits 2 in. ID Tubing
Unit Size:
Each
Product Code. 11709659
Brand: Masterflex™ 3121070
Only null left
Add to basket
Add to basket
Cole-Parmer™ Straight Fitting, Barb x NPT(M), Stainless Steel is durable and provides a secure fit.
  • Stainless steel construction
  • Adapts from an NPT male thread to a hose barb
  • Secure and reliable fit
Specifications
Material Stainless Steel
For Use With Fits 2 in. ID Tubing
Packaging Quantity 1 Pack
Tubing Size Fits 2 in. ID Tubing
Port Size 4-7/16 x 2-7/16in.
Thread Size 2 in. NPT(M)
Width (Metric) 62 mm
Product Type Barbed Fitting