missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 302 HQ, S30430, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Stainless Steel, AISI 302 HQ, S30430, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Tin/Boron/Tantalum/Oxygen/Tungsten/Zirconium/Phosphorus/Nitrogen/Molybdenum/Titanium/Cobalt/Aluminium/Niobium/Vanadium/Carbon/Silicon/Manganese/Copper/Nickel/Chromium/Hydrogen/Arsenic/Lead/Zinc/Calcium/Magnesium/Selenium/Antimony |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tin 0.0017 %/Boron 0.0023 %/Tantalum 0.003 %/Oxygen 0.005 %/Tungsten 0.006 %/Zirconium 0.006 %/Phosphorus 0.009 %/Nitrogen 0.01 %/Molybdenum 0.012 %/Titanium 0.026 %/Cobalt 0.034 %/Aluminium 0.035 %/Niobium 0.053 %/Vanadium 0.055 %/Carbon 0.092 %/Silicon 0.88 %/Manganese 1.93 %/Copper 3.41 %/Nickel 9 %/Chromium 18.15 %/Hydrogen (<0.0010) %/Arsenic (0.001) %/Lead (0.001) %/Zinc (0.001) %/Calcium (0.0017) %/Magnesium (0.002) %/Selenium (0.003) %/Antimony (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction