missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 422, S42200, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Stainless Steel, AISI 422, S42200, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Lead/Zirconium/Boron/Antimony/Titanium/Aluminium/Calcium/Tin/Arsenic/Oxygen/Sulfur/Niobium/Phosphorus/Cobalt/Nitrogen/Copper/Vanadium/Carbon/Silicon/Nickel/Manganese/Molybdenum/Tungsten/Chromium/Iron |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead -0.0017 ppm/Zirconium -0.0017 ppm/Boron 0.0006 %/Antimony 0.0009 %/Titanium 0.0017 %/Aluminium 0.0024 %/Calcium 0.0029 %/Tin 0.0035 %/Arsenic 0.0039 %/Oxygen 0.0073 %/Sulfur 0.0095 %/Niobium 0.013 %/Phosphorus 0.021 %/Cobalt 0.022 %/Nitrogen 0.05 %/Copper 0.126 %/Vanadium 0.21 %/Carbon 0.239 %/Silicon 0.44 %/Nickel 0.616 %/Manganese 0.687 %/Molybdenum 0.97 %/Tungsten 1.03 %/Chromium 12.22 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction