missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ StablCal™ Formazin Turbidity Standard 4000 NTU

Product Code. 12709686
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 12709686

Brand: Hach Lange™ 246149

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Type StablCal Formazin Turbidity Standard
Turbidity 4000 NTU
Volume (Metric) 500 mL
compliance-icons
Product Identifier
  • Formazin Turbidity Standard
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to an approved waste disposal plant.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.