Learn More
IKA™ ST-50-M Gamma Tubes
Product Code. 11846820 Shop All IKA Products
Click to view available options
For Use With (Equipment):
ST-50 Dispersing Tool
Unit Size:
Pack of 10
Product Code. 11846820
Brand: IKA™ 0003701500
Only null left
Add to basket
Add to basket
Specifications
Product Type ST-50-M Tube
Volume Range (Metric) 15 to 50 mL
For Use With (Application) Mixing, stirring, extractions, preparation of soil sample suspensions, solubility testing for medication
For Use With (Equipment) ST-50 Dispersing Tool
Packaging Quantity 10/Pk.