missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-5 Antibody (8e11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42922
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunocytochemistry/Immunofluorescence
SSEA-5 Monoclonal specifically detects SSEA-5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SSEA-5 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| Undifferentiated human embryonic stem cells | |
| 0.1 mL | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 800000000000 | |
| Flow Cytometry 0.0763888888888889, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:1000 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction