missing translation for 'onlineSavingsMsg'
Learn More

SSD114 hydrochloride, MedChemExpress

Product Code. p-200097014
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496607 5 mg 5mg
16490247 50 mg 50mg
16464926 100 mg 100mg
16443888 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496607 Supplier Medchem Express Supplier No. HY103668A
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SSD114 hydrochloride is a novel GABAB receptor positive allosteric modulator.

SSD114 hydrochloride is a novel GABAB receptor positive allosteric modulator.

Specifications

Chemical Name or Material SSD114 hydrochloride
CAS 2319790-02-6
Color White
Purity Grade Notes Research
Molecular Formula C18H21ClF3N3O
Quantity 5 mg
Solubility Information DMSO : 130 mg/mL (335.20 mM; Need ultrasonic) ∣H2O : 2 mg/mL (5.16 mM; ultrasonic and warming and heat to 60°C)
SMILES FC(C1=CC=C(C2=CC(OC)=NC(NC3CCCCC3)=N2)C=C1)(F)F.[H]Cl
Molecular Weight (g/mol) 387.83
Formula Weight 387.83
Percent Purity 98.68%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.