Learn More
SSD114 hydrochloride, MedChemExpress
Product Code. 16496607
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496607 5 mg 5mg
16490247 50 mg 50mg
16464926 100 mg 100mg
16443888 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16496607 Supplier Medchem Express Supplier No. HY103668A
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
MedChemExpress SSD114 hydrochloride is a novel GABAB receptor positive allosteric modulator.
SSD114 hydrochloride is a novel GABAB receptor positive allosteric modulator.
Specifications
Chemical Name or Material SSD114 hydrochloride
CAS 2319790-02-6
Color White
Purity Grade Notes Research
Molecular Formula C18H21ClF3N3O
Quantity 5 mg
Solubility Information DMSO : 130 mg/mL (335.20 mM; Need ultrasonic) ∣H2O : 2 mg/mL (5.16 mM; ultrasonic and warming and heat to 60°C)
SMILES FC(C1=CC=C(C2=CC(OC)=NC(NC3CCCCC3)=N2)C=C1)(F)F.[H]Cl
Molecular Weight (g/mol) 387.83
Formula Weight 387.83
Percent Purity 98.68%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation