Learn More
SSB Mouse anti-Canine, Human, Clone: OTI2C8, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500426S
Description
SSB Monoclonal antibody specifically detects SSB in Canine, Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| SSB | |
| Monoclonal | |
| 0.8 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05455 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 478787, 6741 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| OTI2C8 | |
| Unconjugated | |
| SSB | |
| autoantigen La; La; la autoantigen; La autoantigen homolog; La protein; la ribonucleoprotein; La ribonucleoprotein domain family, member 3; La/SSB; LARP3; lupus La antigen; lupus La protein; Lupus La protein homolog; Lupus LA protein homolog (LA ribonucleoprotein) (LA autoantigen homolog); lupus La protein-like; mRNA for autoantigen; OTTHUMP00000204777; sjoegren syndrome type B antigen; Sjogren syndrome antigen B; Sjogren syndrome antigen B (autoantigen La); small RNA binding exonuclease protection factor La; Ssb; SS-B; SS-B/La protein | |
| Full-length protein expressed in 293T cell transfected with human SSB expression vector | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.