missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRSF2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10715682
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10715682 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price
Product Code. 10715682 Supplier Invitrogen™ Supplier No. PA512402

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, chicken, mouse, porcine and rat based on sequence homology.

The protein encoded by SC35 is a member of the serine/arginine (SR)-rich family of pre-mRNA splicing factors, which constitute part of the spliceosome. Each of these factors contains an RNA recognition motif (RRM) for binding RNA and an RS domain for binding other proteins. The RS domain is rich in serine and arginine residues and facilitates interaction between different SR splicing factors. In addition to being critical for mRNA splicing, the SR proteins have also been shown to be involved in mRNA export from the nucleus and in translation. Two transcript variants encoding the same protein and one non-coding transcript variant have been found for SC35. In addition, a pseudogene of SC35 has been found on chromosome 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SRSF2
Gene Accession No. Q01130
Gene Alias D11Wsu175e; MRF-1; myelin regulatory factor 1; Pr264; Protein PR264; Putative myelin regulatory factor 1; SC35; SC-35; serine and arginine rich splicing factor 2; serine/arginine-rich splicing factor 2; Sfrs10; Sfrs2; SFRS2A; splicing component, 35 kDa; Splicing factor SC35; splicing factor, arginine/serine-rich 2; splicing factor, arginine/serine-rich 2 (SC-35); SR splicing factor 2; SRp30b; Srsf2
Gene Symbols SRSF2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 9-39 amino acids from the N-terminal region of human SFRS2.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6427
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.