missing translation for 'onlineSavingsMsg'
Learn More

SRP9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16672775
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16672775 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16672775 Supplier Invitrogen Supplier No. PA5106339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SRP9 Polyclonal Antibody

SRP9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SRP9
Gene Accession No. P49458, P49962
Gene Alias 9kDa; ALURBP; signal recognition particle 9; signal recognition particle 9 kDa protein; signal recognition particle 9kD; signal recognition particle 9kDa; SRP9
Gene Symbols SRP9
Host Species Rabbit
Immunogen A synthesized peptide derived from human SRP9(Accession P49458), corresponding to amino acid residues L38-E86.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27058, 6726, 690345
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.