Learn More
SRD5A1 293T Cell Overexpression Lysate (Denatured), Abnova
Product Code. 16190962
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16190962 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16190962 Supplier Abnova Supplier No. H00006715T01.100uL
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Western Blotting
Specifications
Cell Line 293T
For Use With (Application) Western Blot
Description SRD5A1 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. BC007033
Molecular Weight (g/mol) 28.6
pH Range 6.8
Plasmid pCMV-SRD5A1 full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-SRD5A1 antibody (H00006715-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
compliance-icons
Product Identifier
  • 100UL SRD5A1 293T Cell Transient Overexpression Ly
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Mercaptoethanol