Learn More
SR-3029, MedChemExpress
Product Code. 16499985
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
Product Code. 16499985
Brand: Medchem Express HY100011
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress SR-3029 is a potent and ATP competitive CK1δ and CK1ε inhibitor, with IC50s of 44 nM and 260 nM, respectively, and Kis of 97 nM for both kinases.
SR-3029 is a potent and ATP competitive CK1δ and CK1ε inhibitor, with IC50s of 44 nM and 260 nM, respectively, and Kis of 97 nM for both kinases.
Specifications
Chemical Name or Material SR-3029
CAS 1454585-06-8
Color White
Purity Grade Notes Research
Molecular Formula C23H19F3N8O
Quantity 2 mg
Solubility Information DMSO : ≥ 30 mg/mL (62.44 mM)
SMILES FC1=CC(N2C=NC3=C(NCC4=NC5=CC=C(F)C(F)=C5N4)N=C(N6CCOCC6)N=C23)=CC=C1
Molecular Weight (g/mol) 480.45
Formula Weight 480.45
Percent Purity 97.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease